Skip to Content
Merck
All Photos(1)

Key Documents

PHL22642

Rotenone

new

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42
Beilstein:
6773081
MDL number:
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

bp

210-220 °C/0.5 mmHg (lit.)

mp

159-164 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat ()

storage temp.

−20°C

SMILES string

COc1cc2OCC3Oc4c5C[C@@H](Oc5ccc4C(=O)C3c2cc1OC)C(C)=C

InChI

1S/C23H22O6/c1-11(2)16-8-14-15(28-16)6-5-12-22(24)21-13-7-18(25-3)19(26-4)9-17(13)27-10-20(21)29-23(12)14/h5-7,9,16,20-21H,1,8,10H2,2-4H3/t16-,20-,21+/m1/s1

InChI key

JUVIOZPCNVVQFO-HBGVWJBISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Inhibitor of mitochondrial electron transport.
Rotenone is an inhibitor of mitochondrial electron transport at nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. It is readily absorbed through the exoskeletons of arthropods, but poorly absorbed cutaneously or from the gastrointestinal tract of mammals. Rotenone acts as a neurotoxic agent which can produce Parkinson-like condition to serve as an animal model for the study of etiology and interventions. The primary mechanism of action of rotenone is complex I inhibition, followed by the generation of oxidative stress and oxidative damage. Because rotenone is lipophilic, it readily penetrates the brain, where it attaches to and inhibits mitochondrial complex I of electron chain transport (ETC).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service