Skip to Content
Merck
Rotenone PESTANAL®, analytical standard

Rotenone

Rotenone PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3