Skip to Content
Merck
All Photos(1)

Key Documents

N12380

Metalaxyl

analytical standard

Synonym(s):

N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-DL-alanine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
Beilstein:
2947777
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1099

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

COCC(=O)N(C(C)C(=O)OC)c1c(C)cccc1C

InChI

1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3

InChI key

ZQEIXNIJLIKNTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alipio Bermúdez-Couso et al.
The Science of the total environment, 461-462, 282-289 (2013-06-07)
The mobility of contaminants in soil is highly dependent upon the characteristics of the contaminant chemical and the properties of the soil. In order to explore these relationships, the district of A Limia (Galicia, NW Spain) was selected as the
Peng Xu et al.
Chemosphere, 83(8), 1074-1079 (2011-02-15)
Knowledge about the enantioselective bioavailability of chiral pesticides in soil invertebrates facilitates more accurate interpretation of their environmental behaviors. In this study, the acute toxicities of R-metalaxyl and rac-metalaxyl to earthworm (Eisenia foetida) were assayed by filter paper contact test.
P Sukul et al.
Reviews of environmental contamination and toxicology, 164, 1-26 (2003-02-18)
Metalaxyl is a systemic fungicide used to control plant diseases caused by Oomycete fungi. Its formulations include granules, wettable powders, dusts, and emulsifiable concentrates. Application may be by foliar or soil incorporation, surface spraying (broadcast or band), drenching, and seed
R Sowmya et al.
Marine biotechnology (New York, N.Y.), 13(5), 918-927 (2011-01-19)
Studies were carried out to utilize in situ proteases of shrimp heads to recover carotenoproteins possessing antioxidant activity. Highest protease activity of the buffer extract was found at pH 8.0 (9.85 ± 0.61 units). The protease activity increased with temperature up to
Fei Ding et al.
Chirality, 24(6), 471-480 (2012-05-01)
Metalaxyl is an acylamine fungicide, belonging to the most widely known member of the amide group. This task is aimed to scrutinize binding region and spatial structural change of principal vector human serum albumin (HSA) complex with (R)-/(S)-metalaxyl by exploiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service