Key Documents
SYNB262910
Synthware bubbler with PTFE stopcock
top joint: ST/NS 29/42, hose size 10 mm
Synonym(s):
Synthware B262910
Select a Size
1 EA
$211.00
$211.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$211.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$211.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
top joint: ST/NS29/42
packaging
pack of 1 ea
manufacturer/tradename
B262910
hose size
10 mm
Related Categories
General description
Synthware bubbler has a small ball joint to prevent mineral oil being sucked back under sudden negative pressure in the system.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)