Skip to Content
Merck
All Photos(1)

Key Documents

CRM69113

Supelco

Cyazofamid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4-Chloro-1-(dimethylaminosulfonyl)-5-(p-tolyl)imidazole-2-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$121.00

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$121.00

About This Item

Empirical Formula (Hill Notation):
C13H13ClN4O2S
CAS Number:
Molecular Weight:
324.79
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CN(C)S(=O)(=O)n1c(nc(Cl)c1-c2ccc(C)cc2)C#N

InChI

1S/C13H13ClN4O2S/c1-9-4-6-10(7-5-9)12-13(14)16-11(8-15)18(12)21(19,20)17(2)3/h4-7H,1-3H3

InChI key

YXKMMRDKEKCERS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Cyazofamid is a foliar and soil fungicide that belongs to the novel chemical class of cyanoimidazole compounds. It is used against various plasmodiophoromycetes and oomycete diseases on fruits and vegetables. It targets the complex III of the enzyme cytochrome bc1 complex in mitochondria and prevents the electron transfer at the Qi (ubiquinone reducing site) center, thereby inhibiting energy supply and respiration.
Cyazofamid is allowed for its use in the European Union as per the Commission Implementing Regulation (EU) 2021/843 of 26th May 2021, renewing its approval as an active substance in plant protection products according to Regulation (EC) No 1107/2009 of the European Parliament and of the Council. Maximum residue levels of cyazofamid allowed for its presence in products of plant and animal origin lie in the range of 0.01-20 mg/kg.
Cyazofamid has to be monitored in the Multiannual Control Program for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Cyazofamid CRM may also find below-given uses:

  • Quantitative assessment of tomato peels for the presence of acephate, chlorpyrifos, and cyazofamid gathered during different intervals of the pre-harvest season by paper spray ionization mass spectrometry (PSI-MS)
  • Multi-residue analysis of 260 pesticides in 74 green leafy vegetable samples with an optimized QuEChERS method for sample preparation and liquid chromatography-tandem mass spectrometry (LC-MS/MS) for detection
  • Simultaneous determination of 116 pesticides from 35 different chemical classes in honey samples by QuEChERS and ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Development of a method for simultaneous quantification of 200 pesticide residues in lettuce and orange samples based on methanol as the extraction solvent, ultra-high-performance liquid chromatography (UHPLC) for separation, and tandem mass spectrometry (MS/MS) for detection

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dissipation kinetics and leaching of cyazofamid fungicide in texturally different agricultural soils
Singh N and Tandon S
International Journal of Environmental Science and Technology, 12(8), 2475-2484 (2015)
V Hanot et al.
Journal of chromatography. A, 1384, 53-66 (2015-02-11)
In 2004, a new multi-residue pesticides method had been published using methanol as extraction solvent. Our goal for this study was to optimize the analytical scheme while extending the compound scope from 19 to 200 pesticides. The main changes from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service