Skip to Content
Merck
All Photos(1)

Key Documents

CRM01345

Supelco

Fluopyram

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-[2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]ethyl]-2-(trifluoromethyl)benzamide, N-{2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]ethyl}-α,α,α-trifluoro-o-toluamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$159.00

$159.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$159.00

About This Item

Empirical Formula (Hill Notation):
C16H11ClF6N2O
CAS Number:
Molecular Weight:
396.71
Beilstein:
11323270
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$159.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

FC(F)(F)c1cnc(CCNC(=O)c2ccccc2C(F)(F)F)c(Cl)c1

InChI

1S/C16H11ClF6N2O/c17-12-7-9(15(18,19)20)8-25-13(12)5-6-24-14(26)10-3-1-2-4-11(10)16(21,22)23/h1-4,7-8H,5-6H2,(H,24,26)

InChI key

KVDJTXBXMWJJEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Fluopyram belongs to the class of succinate dehydrogenase inhibitor (SDHI) fungicides that are also used as a nematicide. It binds to complex II and blocks the mitochondrial electron transport chain, a fundamental component of the Krebs cycle. It is used to control grey mold, white mold (sclerotinia), and powdery mildew in crops such as table grapes, pome fruit, stone fruits, and vegetables, in addition to being used as a seed treatment for the control of nematodes. It is a broad-spectrum systemic fungicide that is absorbed by plant roots and leaves and distributed throughout the plant.
Fluopyram was approved on 1st February 2014 under the Commission Implementing Regulation (EU) No 802/20133 as per Regulation (EC) No 1107/20094 as revised by the Commission Implementing Regulations (EU) No 540/20115 and 541/201. Maximum residue levels (MRLs) for fluopyram have been set according to Reg (EU) 2021/1807 in various products of plant and animal origin from 0.01 to 60 mg/kg. Fluopyram has to be monitored in the Multiannual Control Programmes for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques.
Fluopyram CRM may also find its use as described below:
  • Method development for analyzing the succinate-dehydrogenase-inhibitor fungicide residue levels in vegetable and fruit samples by liquid chromatography-tandem mass spectrometry
  • Development and validation of monoclonal antibody-based immunoassays to analyze fluopyram residues in food samples
  • Simultaneous estimation of fluopyram and tebuconazole in watermelon and soil samples using QuEChERS and GC-MS based method
  • Determination of fluopyram, flupyradifurone, and indaziflam in date fruit, pistachio, and soil samples by ultra-high performance liquid chromatography with tandem mass spectrometry(UHPLC-MS)
  • Develop and validate a method based on QuEChERS and UHPLC-Orbitrap-MS to determine fluopyram and its metabolites in cherry tomato and cucumber

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of Alternative New Pesticide (Fluopyram, Flupyradifurone, and Indaziflam) Residues in Pistachio, Date, and Soil by Liquid Chromatography Triple Quadrupole Tandem Mass Spectrometry
Mahdavi V, et al.
Soil & Sediment Contamination, 373-383 null
Eric Ceballos-Alcantarilla et al.
Food chemistry, 288, 117-126 (2019-03-25)
Monoclonal antibody-based techniques have become a useful analytical technology in the agro-food sector. Nowadays, residues of the recently registered fungicide fluopyram are increasingly being found in quality control programs. In the present study, novel chemical derivatives of this pesticide were
Dissipation and residue determination of fluopyram and tebuconazole residues in watermelon and soil by GC-MS
Dong B and Hu J
International Journal of Environmental Analytical Chemistry, 493-505 null
Antonio Abad-Fuentes et al.
Analytical and bioanalytical chemistry, 407(14), 4207-4211 (2015-03-23)
In recent years, a second generation of succinate-dehydrogenase-inhibitor (SDHI) fungicides has been introduced into the market for effective treatment of fruit and vegetable crops, with fluxapyroxad, boscalid, fluopyram, penflufen, bixafen, penthiopyrad, and isopyrazam being some of the members of this
Marta Vargas-Pérez et al.
Journal of the science of food and agriculture, 100(13), 4826-4833 (2020-06-06)
Fluopyram is a pesticide widely used in tomato and cucumber crops cultivation to control fungal diseases that develop especially in environments with moderate temperatures and high humidity, such as in a greenhouse. The pathway of fluopyram dissipation has been monitored

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service