Skip to Content
Merck
All Photos(1)

Key Documents

CRM67609

Supelco

1,2-Dibromo-3-chloropropane

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

DBCP

Sign Into View Organizational & Contract Pricing

Select a Size

CRM67609-100MG
$89.40

$89.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
CRM67609-100MG
$89.40

About This Item

Linear Formula:
CH2BrCHBrCH2Cl
CAS Number:
Molecular Weight:
236.33
Beilstein:
1732077
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$89.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n/D 1.5542

bp

60-65 °C/7.5 mmHg

density

2.081 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

ClCC(Br)CBr

InChI

1S/C3H5Br2Cl/c4-1-3(5)2-6/h3H,1-2H2

InChI key

WBEJYOJJBDISQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 1B - Repr. 1A - STOT RE 2 Oral

Target Organs

Testes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service