Skip to Content
Merck
All Photos(1)

Key Documents

676713

Sigma-Aldrich

1,2-Dibromo-3-chloropropane

97%

Synonym(s):

DBCP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrCHBrCH2Cl
CAS Number:
Molecular Weight:
236.33
Beilstein:
1732077
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n/D 1.5542

bp

60-65 °C/7.5 mmHg

density

2.081 g/mL at 25 °C

functional group

bromo
chloro

SMILES string

ClCC(Br)CBr

InChI

1S/C3H5Br2Cl/c4-1-3(5)2-6/h3H,1-2H2

InChI key

WBEJYOJJBDISQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dibromo-3-chloropropane can be used to prepare:
  • Tritium radiolabeled dibromochloropropane.[1]
  • L-Glutathione conjugates, which are used to alkylate the DNA to yield N7-guanyl adducts.[2]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 1B - Muta. 1B - Repr. 1A - STOT RE 2 Oral

Target Organs

Testes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and characterization of N7-guanyl adducts derived from 1, 2-dibromo-3-chloropropane
Humphreys WG, et al.
Chemical Research in Toxicology, 4(4), 445-453 (1991)
An improved synthesis of labelled dibromochloropropane
Buchman O, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 18(12), 1713-1719 (1981)
1,2-Dibromo-3-chloropropane.
Report on carcinogens : carcinogen profiles, 12, 134-135 (2011-08-20)
Daniel Albert et al.
Arthritis and rheumatism, 51(4), 656-664 (2004-08-31)
To investigate the possibility that environmental agents contribute to the pathogenesis of Wegener's granulomatosis (WG). We undertook an extensive search for possible environmental exposures by developing a comprehensive questionnaire that was administered by telephone interview to 53 patients with WG
Juraj Lábaj et al.
Nutrition and cancer, 57(2), 209-215 (2007-06-19)
The aim of our investigation was to verify the protective effect of lignin on DNA in rat hepatocytes damaged by 3 different genotoxins attacking DNA in a different manner. Hydrogen peroxide was used for induction of direct single strand breaks

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service