Skip to Content
Merck
All Photos(1)

Key Documents

CRM38373

Supelco

Rimsulfuron

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[3-(ethylsulfonyl)-2-pyridylsulfonyl]urea

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$106.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$106.00

About This Item

Empirical Formula (Hill Notation):
C14H17N5O7S2
CAS Number:
Molecular Weight:
431.44
Beilstein:
7501778
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

CCS(=O)(=O)c1cccnc1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

InChI

1S/C14H17N5O7S2/c1-4-27(21,22)9-6-5-7-15-12(9)28(23,24)19-14(20)18-13-16-10(25-2)8-11(17-13)26-3/h5-8H,4H2,1-3H3,(H2,16,17,18,19,20)

InChI key

MEFOUWRMVYJCQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Rimsulfuron is a sulfonylurea herbicide, used for postemergence use in many crops like potatoes, maize, tomatoes, etc. to control grasses and some broadleaf weeds. Like other sulfonylureas, the site of action of rimsulfuron is acetolactate synthase (ALS)— an enzyme for the biosynthesis of branched-chain amino acids .

As per Regulation (EC) No 1107/2009, repealing the directive 91/414/EEC, rimsulfuron is approved for use in the European Union(EU). A maximum residue limit (MRL) of 0.01-0.02 mg/kg is applicable for the presence of rimsulfuron in fresh/frozen fruits, and tree nuts, in accordance with Regulation Reg. (EU) 617/2014.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

The certified reference material(CRM) rimsulfuron may find the following uses:

  • To study the basis of the selectivity of the herbicide rimsulfuron in maize
  • Quantify leaching of the widely used low-dosage sulfonylurea herbicides rimsulfuron and its primary degradation products
  • Residue analysis of a sulfonylurea herbicide, rimsulfuron, and its dissipation in soil and potato plants under field conditions using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS)
  • Analysis of electrochemical behavior and determination of rimsulfuron herbicide by square wave voltammetry
  • Simultaneous determination of rimsulfuron and haloxyfop-p-methyl and its metabolite haloxyfop in tobacco leaf by liquid chromatography with tandem mass spectrometry (LC-MS/MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service