Skip to Content
Merck
All Photos(1)

Key Documents

18996

Supelco

Metrafenone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(3-Bromo-6-methoxy-2-methylphenyl)(2,3,4-trimethoxy-6-methylphenyl)methanone, 3′-Bromo-2,3,4,6′-tetramethoxy-2′,6-dimethylbenzophenone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$137.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$137.00

About This Item

Empirical Formula (Hill Notation):
C19H21BrO5
CAS Number:
Molecular Weight:
409.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$137.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

SMILES string

COc1ccc(Br)c(C)c1C(=O)c2c(C)cc(OC)c(OC)c2OC

InChI

1S/C19H21BrO5/c1-10-9-14(23-4)18(24-5)19(25-6)15(10)17(21)16-11(2)12(20)7-8-13(16)22-3/h7-9H,1-6H3

InChI key

AMSPWOYQQAWRRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Metrafenone is a systemic fungicide used for its protective, curative, and anti-sporulant properties. It belongs to the chemical class of benzophenones and is used against Pseudocercosporella herpotrichoides and Erysiphe graminis in winter and spring wheat, and barley and Uncinula necator in grapes.
According to the Commission Implementing Regulation (EU) No 540/2011 of 25th May 2011, metrafenone is approved for use in the European Union (EU) as an active substance in plant protection products, following the Regulation (EC) No. 1107/2009. Maximum residue levels (MRLs) of 0.01 mg/kg to 7 mg/kg are set for the presence of metrafenone in products of plant and animal origin, according to the EU Reg. No 2018/687.
Metrafenone has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Metrafenone CRM may also be used in the residue analysis of:
  • 209 pesticides and persistent organic pollutants (POPs), extracted by QuEChERS-based procedure from fresh bovine and chicken liver samples, using gas chromatography coupled to tandem mass spectrometry (GC-MS/MS)
  • 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry
  • 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • 379 pesticides in human serum samples extracted by QuEChERS method using liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Metrafenone in nine fresh vegetable samples using matrix solid-phase dispersion (MSPD) and HPLC-UV
  • Metrafenone in bitter gourd and soil samples using gas chromatography combined with an electron capture detector (GC-ECD)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service