RAD120
radiello™ Diffusive Bodies
white, configured for general use, pk of 20
About This Item
Recommended Products
material
polyethylene body
Quality Level
packaging
pk of 20
manufacturer/tradename
Radiello Code 120
technique(s)
passive air sampling: suitable
thickness × diffusive path length
1.7 mm × 18 mm
color
white
pore size
25 μm average pore size
application(s)
air monitoring
environmental
industrial hygiene
compatibility
configured for general use
Looking for similar products? Visit Product Comparison Guide
General description
Dimensions are 60 mm height x 16 mm diameter. There are four different diffusive bodies available. Each of which are application specific in design.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
radiello® Diffusive Air Sampling Application - Hydrogen Sulfide
Learn about radiello® Diffusive Sampling Aldehydes Applications & Methods.
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocols
radiello® Diffusive Air Sampling Application - Volatile Organic Compounds (VOCs) Chemically Desorbed With CS2
Learn more about radiello air sampler calibration solutions for aldehydes and H2s. We also offer many air sampler calibration kits.
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Related Content
Frequently Asked Questions - radiello® Products
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service