Skip to Content
Merck
All Photos(1)

Key Documents

93981

Supelco

Diol-functionalized Silica gel Chromatography Media

spherical, 200-360 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$461.00
1 KG
$3,110.00

$461.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$461.00
1 KG
$3,110.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
23201100
NACRES:
NB.21

$461.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Diol-functionalized silica gel spherical,

grade

standard

form

powder

composition

carbon content, 4.0-7.0%

technique(s)

LPLC: suitable

surface area

~300 m2/g

matrix

Silica

matrix active group

diol-functionalized silica

particle size

40-75 μm

pore size

110 Å pore size

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT SE 2 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service