Skip to Content
Merck
All Photos(1)

Documents

60757

Supelco

C₁₈-Reversed Phase Silica Gel Chromatography Media

230-400 mesh, fully endcapped

Synonym(s):

Reversed phase silica gel 90 C18

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12000000
NACRES:
SB.52

product name

C18-reversed phase silica gel, for column chromatography, fully endcapped

grade

for column chromatography

Quality Level

form

gel form

quality

fully endcapped

technique(s)

LPLC: suitable

surface area

400 m2/g

surface coverage

max. surface coverage (C) 17‑18% ±4 μmol/m2

matrix

Silica

matrix active group

C18 phase

particle size

230-400 mesh
40-63 μm

pore size

90 Å pore size

separation technique

reversed phase

Related Categories

Application

for reversed phase and ion pair chromatography

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erica C Larson et al.
PloS one, 15(5), e0233485-e0233485 (2020-05-30)
Antimicrobial resistance is a growing global health and economic concern. Current antimicrobial agents are becoming less effective against common bacterial infections. We previously identified pyrrolocins A and C, which showed activity against a variety of Gram-positive bacteria. Structurally similar compounds
Seham S El-Din El-Hawary et al.
Antioxidants (Basel, Switzerland), 8(12) (2019-12-19)
The prevalence of hepatic diseases globally and in Egypt particularly necessitates an intensive search for natural hepatoprotective candidates. Despite the traditional use of Chrysophyllum oliviforme L. and C. cainito L. leaves in the treatment of certain ailments, evidence-based reports on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service