Skip to Content
Merck
All Photos(1)

Documents

807158

TSKgel® SCX (H+) (5 µm) HPLC Column

L x I.D. 30 cm × 7.8 mm, sulfonic acid phase

Synonym(s):

TSKgel® SCX HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® SCX (H+) HPLC Column, phase sulfonic acid, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L22

product line

TSKgel®

feature

counter ion H+

manufacturer/tradename

Tosoh 07158

parameter

≤20% organic solvent
1.2 mL/min flow rate
10-45 °C temperature
50 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polystyrene-divinylbenzene

matrix active group

sulfonic acid phase

particle size

5 μm

operating pH

1-14

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

This TSKgel® SCX column is packed with 5μm porous spherical particles which are surface modified with a strong (sulfonic acid) cation exchanger in the hydrogen. The column is optimized for the separation and analysis of organic acids, saccharides and alcohols. TSKgel SCX (H+) columns are mainly applied to the analysis of isomerized sugars, alcohols and lower organic acids.

Physical form

Shipped in DI water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service