Skip to Content
Merck
All Photos(1)

Key Documents

77184

Supelco

Elemental Impurities according to ICH Q3D parenteral, Standard 5

TraceCERT®, (in 12% Nitric acid), applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

Agency

according to ICH Q3D
according to Ph. Eur.
according to USP

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

Ag, 15 mg/L
As, 15 mg/L
Cd, 2 mg/L
Co, 5 mg/L
Hg, 3 mg/L
Ni, 20 mg/L
Pb, 5 mg/L
Se, 80 mg/L
Ti, 8 mg/L
V, 10 mg/L

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

(in 12% Nitric acid)

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the certificate.

Download your certificate at: https://www.sigmaaldrich.com/US/en/documents-search

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation. 

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Other Notes

Please visit Inorganic Elemental Impurity Mix Standards for a complete product listing of Elemental Impurity Mixes and Single Element Certified Reference Material Solutions

For detailed information for on our newly launched Elemental Impurity Mixes please check the flyer: Elemental Impurities - Certified Reference Materials for ICH Q3D, USP <232> and Ph.Eur. 5.20

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service