Skip to Content
Merck
All Photos(1)

Key Documents

19041

Supelco

Elemental Impurities according to ICH Q3D oral, Standard 1

TraceCERT®, (in 12% nitric acid), applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20

Synonym(s):

TraceCERT Elemental Impurities according to ICH Q3D oral

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

according to ICH Q3D
according to Ph. Eur.
according to USP

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

Ag, 150 mg/L
As, 15 mg/L
Cd, 5 mg/L
Co, 50 mg/L
Hg, 30 mg/L
Ni, 200 mg/L
Pb, 5 mg/L
Se, 150 mg/L
Tl, 8 mg/L
V, 100 mg/L

concentration

(in 12% nitric acid)

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Other Notes

We provide a diverse range of certified reference materials, including various elemental impurity mixes and single elements, with concentration limits that comply with ICH Q3D guidelines. For more information please visit our Inorganic Elemental Impurity Mix Standards.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service