Skip to Content
Merck
All Photos(1)

Documents

58696-U

PTFE Tubing

L × O.D. × I.D. 50 ft × 1/16 in. × 0.031 in.

Synonym(s):

Polytetrafluoroethylene Hose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32119290
NACRES:
SB.52

parameter

500 psi max. pressure

L × O.D. × I.D.

50 ft × 1/16 in. × 0.031 in.

Looking for similar products? Visit Product Comparison Guide

General description

Economical, flexible PTFE tubing is ideal for use at pressures up to 500 psi (35 kg/cm2). Use in automatic analyzer, postcolumn reaction, preparative scale systems, in-stream sampling devices, and when monitoring physiologically important compounds.

The maximum recommended operating temperature for PTFE is 200 °C, but short term exposure to higher temperatures seldom causes damage.

Application


  • How changes in column geometry and packing ratio can increase sample load and throughput by a factor of fifty in Counter-Current Chromatography.: This research discusses the optimization of column geometry and packing ratios in counter-current chromatography, enhancing sample load capacity and throughput using PTFE tubing, which serves as a flexible and chemically inert medium for the operation (Peng et al., 2018).

  • Sequential injection anodic stripping voltammetry with monosegmented flow and in-line UV digestion for determination of Zn(II), Cd(II), Pb(II) and Cu(II) in water samples.: This study showcases a method using PTFE tubing in a sequential injection system to analyze heavy metals in water samples, emphasizing the tubing′s resistance to corrosive conditions and its role in sample transport (Siriangkhawut et al., 2011).

  • Photo-induced fluorescence of magnesium derivatives of tetracycline antibiotics in wastewater samples.: This article explores the use of PTFE tubing in a setup for detecting antibiotics in wastewater through photo-induced fluorescence, illustrating PTFE′s utility in maintaining sample integrity under photochemical conditions (Pena et al., 2010).

  • Online phototransformation-flow injection chemiluminescence determination of triclosan.: Research on the online photochemical transformation of triclosan, utilizing PTFE tubing in the chemiluminescence detection system, highlights its effectiveness in sample handling and reaction containment (Song et al., 2007).

  • High-performance liquid chromatography: photochemical reduction in aerobic conditions for determination of K vitamins using fluorescence detection.: The study employs PTFE tubing in an HPLC system for the photochemical reduction of vitamins, demonstrating the tubing′s role in enhancing detection sensitivity and preventing sample degradation (Pérez-Ruiz et al., 2007).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service