Skip to Content
Merck
All Photos(1)

Documents

57266

Supelco

Visiprep Manifold Cover (flow control valves and gasket included)

for use with DL (Disposable Liner), 24-port model

Synonym(s):

TOP PLATE W/FLOW CONTROL VALVES, 24 PORT VISIPREP-DL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with DL (Disposable Liner), 24-port model

Looking for similar products? Visit Product Comparison Guide

Footnote

Included with Visiprep DL 24-port Vacuum Manifold (57265)

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service