57160-U
Preppy 12-Port Vacuum Manifold
Synonym(s):
12 port SPE cartidge tube vacuum manifold
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product line
Preppy
Quality Level
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
The Preppy manifold is our simplest and most economical manifold. It too enables the analyst to simultaneously prepare up to 12 samples. It consists of a chemical-resistant cover and gasket, a glass basin, a vacuum release vent, 12 individual control valves with knurled tops, and stainless steel solvent guide needles.
Two optional collection racks are available; one holds both 2 and 4 mL autosampler vials, and the other holds 15 and 20 mL vials. An optional vacuum gauge/bleed valve assembly can be installed to allow precise control of the vacuum used with the Preppy manifold.
Two optional collection racks are available; one holds both 2 and 4 mL autosampler vials, and the other holds 15 and 20 mL vials. An optional vacuum gauge/bleed valve assembly can be installed to allow precise control of the vacuum used with the Preppy manifold.
Legal Information
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service