Skip to Content
Merck

Skip To

VM20

VM20 Vacuum Manifold

For processing up to 20 plasmid purification samples simultaneously

Synonym(s):

Vacuum Assembly

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 set
Check Cart for Availability
$262.00

About This Item

NACRES:
NA.52
UNSPSC Code:
41105501

$262.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


General description

The VM20 Vacuum Manifold is designed to provide increased efficiency and performance when used with many of Sigma′s column-based nucleic acid purification systems. The manifold is capable of processing 20 samples simultaneously and includes a stepped design for easy access to every column. Dual vacuum connection ports allow for multiple configurations for waste collection. Waste can be collected in the manifold body or, through use of a threaded drain port, diverted to any auxiliary reservoir providing virtually unlimited capacity. The manifold includes chemically resistant Endura-Luer stopcocks that will not crack or break with repeated use.
This product is made out of chemically resistant polypropylene.
We have created Endura-Luers to be an enhanced stopcock that
resists chemical erosion, cracking, and breaking.

Application

VM20 Vacuum Manifold has been used for the isolation of circulating tumor cells by filtration with track-etched membranes.[1]

Features and Benefits

  • Capable of processing 20 samples simultaneously
  • The dual port design allows waste to be collected in the manifold body or diverted to any auxiliary reservoir
  • Stepped design for easy access to every column
  • Endura-Luer stopcocks instead of plugs
  • Chemically resistant polypropylene

Other Notes

GenElute HP Plasmid Miniprep Kits
GenElute Five-Minute Plasmid Miniprep Kits
GenElute HP Plasmid Midiprep Kit
GenElute HP Plasmid Maxiprep Kit
HIS-Select® Cartridge
HIS-Select® High Flow Cartridge (1.2 ml capacity)
HIS-Select® High Flow Cartridge (6.4 ml capacity)
Kit contains the Sigma VM20 Vacuum manifold, 20 Endura-Luer stopcocks, and VM20 Accessory Pack with cap and 2 nozzles.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC
HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

A step-by-step protocol for released N-linked glycan analysis of the monoclonal antibody adalimumab, based on UHPLC-FLR-MS and procainamide labeling.

Follow this simple, rapid, and cost-effective method for isolating plasmid DNA from recombinant E. coli cultures with the GenElute™ HP Plasmid Maxiprep kit.

Related Content


T G Ntouroupi et al.
British journal of cancer, 99(5), 789-795 (2008-08-07)
We have developed an automated, highly sensitive and specific method for identifying and enumerating circulating tumour cells (CTCs) in the blood. Blood samples from 10 prostate, 25 colorectal and 4 ovarian cancer patients were analysed. Eleven healthy donors and seven



Global Trade Item Number

SKUGTIN
VM20-1SET04061837481697

Questions

Reviews

No rating value

Active Filters