57148-U
SPE tube
Si-5G-44%H2SO4/Si SPE Tube, volume 12mL, pk of 20
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
12mL Si-5G-44%H2SO4/Si SPE Tube, pk of 20
material
polypropylene tube
composition
bed wt., 5 g
packaging
pk of 20
technique(s)
solid phase extraction (SPE): suitable
matrix active group
silica bonding
application(s)
food and beverages
storage temp.
2-8°C
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service