Skip to Content
Merck
All Photos(1)

Documents

21334-U

Supelco

Silica Gel

44% H₂SO₄ Coated, pkg of 100 g bulk packing

Synonym(s):

Sulfuric Acid Coated Silica Gel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NB.21

product name

44% H2SO4 Coated Silica Gel, pkg of 100 g bulk packing

Agency

suitable for EPA 1613

form

solid

packaging

pkg of 100 g bulk packing

technique(s)

solid phase extraction (SPE): suitable

particle size

60-200 mesh

application(s)

food and beverages

General description

The treated silica gels that may be primarily employed in the pre-packed multi-layer silica gel columns are available in bulk packages. These are attractive options for customizing the required columns to more efficiently clean very dirty samples, or to prepare shorter columns when samples are relatively clean, e.g. drinking water.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service