Skip to Content
Merck
All Photos(1)

Key Documents

47839

Supelco

Sodium Saccharin

analytical standard

Synonym(s):

Saccharin sodium salt hydrate, 2,3-Dihydro-3-oxobenzisosulfonazole sodium salt, 2-Sulfobenzoic acid imide sodium salt, o-Sulfobenzimide sodium salt, Saccharin soluble

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4NNaO3S · xH2O
CAS Number:
Molecular Weight:
205.17 (anhydrous basis)
Beilstein:
3599229
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E954
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

[Na+].[H]O[H].[O-]C1=NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S.Na.H2O/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;;/h1-4H,(H,8,9);;1H2/q;+1;/p-1

InChI key

ILJOYZVVZZFIKA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium saccharin is a white crystalline powder[1], which is commonly used as a synthetic or artificial sweetener in carbonated soft drinks and beverages.[2] It can find a wide range of applications in food processing, commodities and also feed production industries.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Encyclopedia of Food and Color Additives, 1 (1997)
Encyclopedia of Food and Color Additives, 1 (1997)
Rapid and sensitive detection of sodium saccharin in soft drinks by silver nanorod array SERS substrates
Han C, et al.
Sensors and Actuators B, Chemical, 272-279 (2017)
Heather E King et al.
Pharmacology, biochemistry, and behavior, 126, 163-169 (2014-10-07)
The inbred Fischer (F344) and Lewis (LEW) rats, while originally developed as animal models for cancer and tissue transplantation research, have since been used to study genetic differences in a variety of physiological and behavioral endpoints. In this context, LEW
Steven Zukerman et al.
Chemical senses, 34(8), 685-694 (2009-09-09)
Prior work has shown that sweet taste-deficient T1R3 knockout (KO) mice developed significant sucrose preferences when given long-term sugar versus water tests. The current study investigated the role of olfaction in this experience-conditioned sucrose preference. T1R3 KO and C57BL/6 wild-type

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service