Skip to Content
Merck
All Photos(3)

Key Documents

240931

Sigma-Aldrich

Saccharin

≥99%

Synonym(s):

2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$66.50
50 G
$168.00

$66.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$66.50
50 G
$168.00

About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
Beilstein:
6888
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$66.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

powder

mp

226-229 °C (lit.)

solubility

acetone: 1g in 12mL(lit.)
alcohol: 1g in 31mL(lit.)
boiling water: 1g in 25mL(lit.)
water: 1g in 25mL(lit.)

SMILES string

O=C1NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)

InChI key

CVHZOJJKTDOEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The biodegradation of saccharin (2-sulfobenzoic acid imide), an artificial sweetening agent, was studied[1]. It was also studied for its carcinogenic potential.

Application


  • Gabapentin-saccharin co-crystals with enhanced physicochemical properties and in vivo absorption formulated as oro-dispersible tablets.: This study presents the formulation of gabapentin-saccharin co-crystals, demonstrating improved solubility and absorption, beneficial for pharmaceutical applications (Soliman et al., 2020, doi: 10.1080/10837450.2019.1687521).

  • Improving the solubility, dissolution, and bioavailability of Ibrutinib by preparing it in a coamorphous state with saccharin.: The research focuses on enhancing the bioavailability of Ibrutinib through coamorphous preparation with saccharin, which is significant for cancer treatment (Shi et al., 2019, doi: 10.1016/j.xphs.2019.04.031).

Biochem/physiol Actions

A sweet tastant for mammals. A glycerol taste receptor binding site specific for glucose has been proposed in drosophila.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N H Tran et al.
Bioresource technology, 161, 40-46 (2014-04-01)
The biodegradation of the six artificial sweetening agents including acesulfame (ACE), aspartame (ASP), cyclamate (CYC), neohesperidindihydrochalcone (NHDC), saccharin (SAC), and sucralose (SUC) by nitrifying activated sludge was first examined. Experimental results showed that ASP and NHDC were the most easily
Hellmuth Lilienthal et al.
Toxicology, 311(1-2), 52-60 (2013-02-14)
Many chemicals are known to exhibit endocrine activity and affect reproductive functions in vertebrates and invertebrates. Endocrine effects include influences on sexual differentiation of the brain during development and reproductive and non-reproductive behavior in adult offspring. We previously demonstrated that
Katharine J Tuerke et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(40), 13709-13717 (2012-10-05)
A taste associated with emetic drugs produces conditioned disgust reactions in rats (predominantly gaping), unlike nonemetic drugs that can still produce conditioned taste avoidance but not conditioned disgust. That difference suggests nausea is a prerequisite for learning disgust reactions to
Rino Ragno et al.
Journal of medicinal chemistry, 50(6), 1241-1253 (2007-02-28)
The screening of the inhibition capabilities of dye-like small molecules from a focused library against both human PRMT1 and Aspergillus nidulans RmtA is reported as well as molecular modeling studies (homology modeling, molecular docking, and 3-D QSAR) of the catalytic
L B Ellwein et al.
Critical reviews in toxicology, 20(5), 311-326 (1990-01-01)
Almost from its discovery in 1879, the use of saccharin as an artificial, non-nutritive sweetener has been the center of several controversies regarding potential toxic effects, most recently focusing on the urinary bladder carcinogenicity of sodium saccharin in rats when

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service