Skip to Content
Merck
All Photos(1)

Key Documents

47178-U

Supelco

Hexaldehyde-2,4-DNPH solution

1000 μg/mL in acetonitrile (as aldehyde), analytical standard

Synonym(s):

Hexanal 2,4-dinitrophenylhydrazone solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetonitrile (as aldehyde)

storage temp.

2-8°C

InChI

1S/C12H16N4O4/c1-2-3-4-5-8-13-14-11-7-6-10(15(17)18)9-12(11)16(19)20/h6-9,14H,2-5H2,1H3/b13-8+

InChI key

SJUYYAXVFFVQNX-MDWZMJQESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product is also available as a certified reference material. Please reference CRM47178.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service