Skip to Content
Merck
All Photos(1)

Key Documents

488127

Sigma-Aldrich

Benzaldehyde-α-13C

99 atom % 13C, 99% (CP)

Synonym(s):

13C Labeled benzaldehyde, Benzaldehyde-(carbonyl-13C), Bitter almond-(carbonyl-13C)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$2,050.00

$2,050.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

Select a Size

Change View
500 MG
$2,050.00

About This Item

Linear Formula:
C6H513CHO
CAS Number:
Molecular Weight:
107.11
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

$2,050.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

refractive index

n20/D 1.545 (lit.)

bp

178-179 °C (lit.)

mp

−26 °C (lit.)

density

1.055 g/mL at 25 °C

mass shift

M+1

SMILES string

[H][13C](=O)c1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H/i6+1

InChI key

HUMNYLRZRPPJDN-PTQBSOBMSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.0 °F - closed cup

Flash Point(C)

62.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service