Skip to Content
Merck
All Photos(1)

Key Documents

46903

Supelco

trans-9-Octadecenoic methyl ester

10 mg/mL in heptane, analytical standard

Synonym(s):

Methyl elaidate, Elaidic acid methyl ester, Methyl trans-9-octadecenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOCH3
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727038
EC Number:
MDL number:
UNSPSC Code:
85151701

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

packaging

ampule of 1 mL

concentration

10 mg/mL in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.871 g/mL at 20 °C (lit.)

format

single component solution

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+

InChI key

QYDYPVFESGNLHU-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farshad Darvishi et al.
New biotechnology, 28(6), 756-760 (2011-02-18)
The yeast Yarrowia lipolytica degrades efficiently low-cost hydrophobic substrates for the production of various added-value products such as lipases. To obtain yeast strains producing high levels of extracellular lipase, Y. lipolytica DSM3286 was subjected to mutation using ethyl methanesulfonate (EMS)
Renee M Thomas et al.
Journal of the American Chemical Society, 133(19), 7490-7496 (2011-04-23)
N-Aryl,N-alkyl N-heterocyclic carbene (NHC) ruthenium metathesis catalysts are highly selective toward the ethenolysis of methyl oleate, giving selectivity as high as 95% for the kinetic ethenolysis products over the thermodynamic self-metathesis products. The examples described herein represent some of the
Cyril Thurier et al.
ChemSusChem, 1(1-2), 118-122 (2008-07-09)
Unsaturated vegetable oils are an attractive renewable feedstock, and the selective cleavage of unsaturated fatty esters is an important transformation in this respect. The efficient and selective cross-metathesis of methyl oleate with ethylene was achieved under mild conditions with ruthenium-alkylidene
Chinmay A Deshmane et al.
Chemical communications (Cambridge, England), 46(34), 6347-6349 (2010-08-18)
Thermally stable mesoporous Ga-Nb mixed oxides, active in both acid-catalysed and redox reactions have been synthesized via self-assembly hydrothermal assisted approach. Methyl oleate, a major component of biodiesels, undergoes double bond and skeletal isomerisation as well as dehydrogenation over these
Atanu Biswas et al.
Journal of agricultural and food chemistry, 56(14), 5611-5616 (2008-06-19)
An environmentally friendly water-based pathway to form the azide derivatives of soybean oil and fatty esters is reported. This entails first the formation of epoxides and then the azidization of the epoxides. The azidization reaction is carried out at high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service