Skip to Content
Merck
All Photos(1)

Key Documents

442666-U

Supelco

n-Butyl acetate

analytical standard

Synonym(s):

Butyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

4 (vs air)

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

790 °F

expl. lim.

7.6 %

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.394 (lit.)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Butyl acetate is a limpid colorless liquid with a fruity odor. It is prepared by heating and distillation of n-butyl alcohol and acetic acid, in presence of sulphuric acid which acts as a catalyst. It is generally used as a solvent, during the production of lacquers, natural gums and synthetic resins.[1]

Application

Butyl acetate is suitable to be used as an organic solvent, for the extraction of phosphomolybdic acid (PMA), which is used for the measurement of inorganic orthophosphate (Pi).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of inorganic orthophosphate in biological materials: extraction properties of butyl acetate.
Sanui H.
Analytical Biochemistry, 60 (2), 489-504 (1974)
Encyclopedic Dictionary of Polymers.
Gooch JW.
Science, 101-101 (2011)
Andréa B Martins et al.
Ultrasonics sonochemistry, 20(5), 1155-1160 (2013-03-05)
The influence of low-frequency ultrasound (40 kHz) in the esterification reaction between acetic acid and butanol for flavor ester synthesis catalyzed by the commercial immobilized lipase B from Candida antarctica (Novozym 435) was evaluated. A central composite design and the
Matias Rauma et al.
Journal of pharmaceutical sciences, 98(11), 4365-4375 (2009-03-14)
Recent work in our laboratory shows that it is possible to reproducibly measure the weight change over time of a piece of porcine skin exposed to a chemical vapor by thermogravimetric analysis (TGA), that is, using a microbalance maintained at
Prateek Kathel et al.
ISA transactions, 49(1), 130-137 (2009-10-10)
This work deals with the dynamics and control of a high-purity batch distillation column with chemical reaction. A heterogeneous esterification reaction between the acetic acid and butanol takes place to produce butyl acetate. The process model is formulated considering variable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service