Skip to Content
Merck
All Photos(2)

Documents

242594

Sigma-Aldrich

sec-Butyl acetate

99%

Synonym(s):

(±)-2-Butyl acetate, (±)-sec-Butyl acetate, 1-Methylpropyl acetate, 2-Acetoxybutane, 2-Butyl acetate, Acetic acid sec-butyl ester, Ultrasolve M 1200, sec-Butyl alcohol acetate (6CI), sec-Butyl ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(CH3)C2H5
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4 (vs air)

Assay

99%

form

liquid

expl. lim.

7.5 %

refractive index

n20/D 1.389 (lit.)

bp

111-112 °C (lit.)

mp

−99 °C (lit.)

solubility

alcohol: soluble(lit.)
diethyl ether: soluble(lit.)
water: slightly soluble(lit.)

density

0.872 g/mL at 25 °C (lit.)

SMILES string

CCC(C)OC(C)=O

InChI

1S/C6H12O2/c1-4-5(2)8-6(3)7/h5H,4H2,1-3H3

InChI key

DCKVNWZUADLDEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronika Ruzsanyi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 911, 84-92 (2012-12-12)
Volatile organic compounds (VOCs) released by humans through their skin were investigated in near real time using ion mobility spectrometry after gas chromatographic separation with a short multi-capillary column. VOCs typically found in a small nitrogen flow covering the skin
Yilong Wang et al.
Chirality (2018-06-10)
We previously identified and characterized 1 novel deep-sea microbial esterase PHE21 and used PHE21 as a green biocatalyst to generate chiral ethyl (S)-3-hydroxybutyrate, 1 key chiral chemical, with high enantiomeric excess and yield through kinetic resolution. Herein, we further explored
W S Wu et al.
The Analyst, 122(4), 321-323 (1997-04-01)
The application of tryptamine to derivatize airborne isocyanates has been evaluated and adopted by the National Institute for Occupational Safety and Health (NIOSH) as the latest isocyanates regulatory method (Method 5522) in the USA. Method 5522 uses dimethyl sulfoxide (DMSO)
Renata Medinaceli Quintela et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(38), 7269-7285 (2020-08-21)
Processing of olfactory information is modulated by centrifugal projections from cortical areas, yet their behavioral relevance and underlying neural mechanisms remain unclear in most cases. The anterior olfactory nucleus (AON) is part of the olfactory cortex, and its extensive connections
Rocío Ríos-Reina et al.
Food research international (Ottawa, Ont.), 123, 298-310 (2019-07-10)
The characteristic volatile profile of the Spanish Vinagre de Jerez (VJ), Vinagre de Condado de Huelva (VC) and Vinagre de Montilla-Moriles (VMM) protected designation of origin (PDO) wine vinegars has been studied and compared for the first time by headspace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service