Skip to Content
Merck
All Photos(1)

Key Documents

442285

Supelco

2,3,5-Trichlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl3O
CAS Number:
Molecular Weight:
197.45
Beilstein:
2046862
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Oc1cc(Cl)cc(Cl)c1Cl

InChI

1S/C6H3Cl3O/c7-3-1-4(8)6(9)5(10)2-3/h1-2,10H

InChI key

WWGQHTJIFOQAOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5-Trichlorophenol belongs to the class of substituted phenol compounds. They are widely used in industrial applications such as synthesis of pesticides, dyes, drugs, plastics, etc. They are found in the environment as pollutant due to their extensive usage.

Application

2,3,5-Trichlorophenol may be used as an analytical reference standard for the determination of the analyte in landfill leachates, aqueous samples, and environmental samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

P T Charles et al.
Bioconjugate chemistry, 6(6), 691-694 (1995-11-01)
A synthetic scheme has been developed for the preparation of a dye-labeled analog of polychlorinated biphenyls. The reaction of 2,3,5-trichlorophenol with 3-bromopropylamine hydrobromide under basic conditions was used to introduce a free primary amine group into the parent compound by
L Drummond et al.
British journal of industrial medicine, 45(7), 493-497 (1988-07-01)
Plasma gamma-hexachlorocyclohexane (gamma-HCH) and three urinary trichlorophenols were measured in forestry workers who were engaged in planting seedlings treated with gamma-HCH. These two procedures were assessed as potential biological monitoring methods and the data were compared with reported clinical symptoms.
V S Bondar et al.
FEMS microbiology letters, 181(1), 73-82 (1999-11-24)
The regiospecificity of hydroxylation of C2-halogenated phenols by Rhodococcus opacus 1G was investigated. Oxidative defluorination at the C2 position ortho with respect to the hydroxyl moiety was preferred over hydroxylation at the non-fluorinated C6 position for all 2-fluorophenol compounds studied.
Evaluation of a dedicated gas chromatography-mass spectrometry method for the analysis of phenols in water.
Vermeulen A, et al.
Journal of Chromatography A, 1071(1-2), 41-46 (2005)
Development and optimization of a method for the analysis of phenols and chlorophenols from aqueous samples by gas chromatography-mass spectrometry, after solid-phase extraction and trimethylsilylation.
Kovacs A, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 99(1), 125-131 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service