Recommended Products
parameter
0.5-3.0 L/min flow rate
32-113 °F temp. range (0-45 °C)
technique(s)
active air sampling: suitable
battery power
4.8 V, 14 hr life
application(s)
air monitoring
environmental
industrial hygiene
Related Categories
General description
Specifications | |
---|---|
Operating Flow Control | Volumetric flow rate held within ± 2.5% of set-point over the 1 to 3 L/min operating range (± 5% to 0.5 LPM), with automatic compenstation for battery voltage, altitude, temperature, and sample load changes. |
Flow Rate | Flow rate adjustable between 0.5 and 3.0 L/min; lower flow rates attainable (to 1 mL/min) with the optional Gemini Twin-Port Sampling accessory |
Flow Indication | Liquid crystal display (LCD) provides 0.01 L/min resolution |
Operating Range | 30 inches of water load upt to 2 L/min and 10 inches up to 3 L/min |
Flow Blockage Detection | Flow-fault LED comes on immedieatly when block is detected. Pump shuts down within 90 seconds if block is not cleared. |
Elapsed Time Readout | To 999 minutes in increments of 1 minute. Holds the last reading after either flow blockage or low battery shutdown and when pump is OFF or in HOLD mode. |
Operating Time | Varies with flow rate and sample device loading. |
Operating Temperature Limits | 32-113 °F (0-45 °C) |
Physical Weight | 23 oz. (652 g) with battery pack |
Dimensions | 2.25 in. D x 4.25 in. H x 4.0 in. W (5.7 cm D x 10.8 cm H x 10.2 cm) with battery pack |
Power Supply | 4.8 V battery pack of four NiMH cells (units mfg before October 2011 contain NiCd cells) |
Battery Pack Capacity | 2.8 amp hours (NiMH version); 2.4 amp hours (NiCD version) |
Battery Pack Recharge Time | 14-16 hours overnight with Omega Battery Charger |
Typical Battery Pack Life | 300 or more charging cycles |
Features and Benefits
- Patented electronic laminar flow control
- Internal secondary standard calibrates the pump continuously, maintaining OSHA compliance
- Check against a primary standard only once a month or every 200 hours for coal mine dust sampling
- Inlet filter traps moisture and particular matter
- Stainless steel-filled plastic case provides EMI/RFI protection
- Rated IP66 for resistance to water and dust ingress
- Exceptionally quiet operation
- Flow-Fault Indicator shows any sample inlet blockage
- Run/Hold feature allows "time out" during sampling process
- Elapsed time readout and low-battery warning
- Low flow and dual tube sampling available with Gemini Twin-Port Sampler (28118-U)
Suitability
- NIOSH certified for coal mine dust sampling (TC-74-030)
- MSHA certified as intrinsically safe for underground use (Approval # 2G-3924-1).
- UL approved as intrinsically safe for use in hazardous locations, Class I, Groups A, B, C, D; Class II, Groups E, F and G; Class III, Division I locations.
Legal Information
recommended
related product
required but not provided
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.
Protocols
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service