27111
Vials, screw top, clear glass (vial only)
volume 4 mL, clear glass vial, thread for 13-425, O.D. × H × I.D. 15 mm × 45 mm × 8 mm, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear glass vial
feature
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
15 mm × 45 mm × 8 mm
volume
4 mL
fitting
thread for 13-425
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
It has been used in vapour trap for determining atmospheric water vapour for isotopic analysis. It was also used in solid phase microextraction (SPME) combined with gas chromatography/mass spectrometry (GC/MS) method during the determination of the oxidation of freeze-dried chicken myofibrils spiked with methyl linoleate.
Suitability
Autosampler compatibility:
Agilent 7673A, Series I, II
Shimadzu LC Autosamplers
Waters 48, 717, 717 Plus
Agilent 7673A, Series I, II
Shimadzu LC Autosamplers
Waters 48, 717, 717 Plus
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Solid phase microextraction- gas chromatography for quantifying headspace hexanal above freeze-dried chicken myofibrils.
Journal of Agricultural and Food Chemistry, 51.15, 4185-4190 (2003)
A rapid method for the sampling of atmospheric water vapour for isotopic analysis.
Rapid Communications in Mass Spectrometry, 24.1, 103-108 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service