Skip to Content
Merck
All Photos(1)

Documents

24818-U

Supelco

Merlin Microseal System Replacement Septum

1 SPME septum

Synonym(s):

GC accessory, SPME septum, Merlin Microseal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.54

General description

The Merlin Microseal System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge "blunt tipped" needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.

Application

Merlin Microseal System may be used to replace traditional septum to avoid leaking and bleeding caused during operating GC/MS system. It may also be equipped to the injection port during GC analysis for determination of plasticizer N-butylbenzenesulfonamide and the pharmaceutical Ibuprofen in wastewater.

Other Notes

Three septa versions are available:
  • Low Pressure for use with 23 gauge syringe needles, and injection port pressures between 1 and 45 psi. Do not use with syringe needles that have beveled tips.
  • General Purpose for use with 23 gauge syringe needles, and injection port pressures between 3 and 100 psi. Do not use with syringe needles that have beveled tips.
  • SPME for use with 23 gauge SPME fiber assemblies. Do not use with SPME fiber assemblies that have beveled tips.

Note: Do not use with beveled tips.

Legal Information

MicroSeal is a trademark of Merlin Instrument Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the plasticizer N-butylbenzenesulfonamide and the pharmaceutical Ibuprofen in wastewater using solid phase microextraction (SPME).
Huppert N
Fresenius Journal of Analytical Chemistry, 326 (6), 529-536 (1998)
J M Diserens
Journal of AOAC International, 84(3), 853-860 (2001-06-22)
Although the use of pentachlorophenol (PCP) is prohibited in most countries, it is still widely found in the wood of pallets, containers, crates and in cardboard, paper, etc. Such material may contain not only PCP but other chlorophenols as well.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service