Skip to Content
Merck
All Photos(8)

Documents

ZRB2124

Sigma-Aldrich

Anti-TREM-2 Antibody, clone 1I11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Triggering receptor expressed on monocytes 2, Triggering receptor expressed on myeloid cells 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1I11
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 25 kDa
observed mol wt ~36-50 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TREM2(54209)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I11 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects TREM2. It targets an epitope within 14 amino acids from the extracellular domain within the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the extracellular domain within the C-terminal half of human TREM2.

Application

Quality Control Testing

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected TREM-2 in RAW264.7 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TREM-2 in lysate from Jurkat cells and 1:10,000 dilution detected TREM-2 in lysate from Human brain tissue.

Flow Cytometry Analysis: 1 µg from a representative lot detected TREM-2 in one million Jurkat cells.

Affinity Binding Assay: A representative lot of this antibody bound TREM-2 peptide with a KD of 2.0 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TREM-2 in THP-1 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Triggering receptor expressed on myeloid cells 2 (UniProt: Q9NZC2; also known as TREM-2, Triggering receptor expressed on monocytes 2) is encoded by the TREM2 gene (Gene ID: 54209) in human. TREM-2 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 19-174), a transmembrane domain (aa 175-195), and a cytoplasmic domain (aa 196-230). TREM-2 is an Ig-like V-type receptor expressed by populations of myeloid cells in various region of the central nervous system. In the CNS strongest expression seen in the basal ganglia, corpus callosum, medulla oblongata and spinal cord. Its expression is also reported on macrophages and dendritic cells, but not on granulocytes or monocytes. TREM-2 is highly expressed in microglia, where it, along with its adaptor protein DAP12, regulates inflammatory cytokine release and phagocytosis of apoptotic neurons. It plays a role in chronic inflammations and may stimulate production of constitutive rather than inflammatory chemokines and cytokines. Its short cytoplasmic tail does not contain any signaling motifs and relies on its association with the immunoreceptor tyrosine-based activation motif- (ITAM-) containing TYROBP for signal transduction. It forms a receptor signaling complex with TYROBP and triggers activation of the immune responses in macrophages and dendritic cells. Apolipoprotein E (ApoE) is reported to function as a high-affinity ligand for TREM-2. ApoE binding is shown to enhance the phagocytosis of apoptotic Neuro2a cells by primary microglia cells. Greatly reduced ApoE binding is seen with Alzheimer s disease-associated TREM-2 R47H mutant. TREM2 gene mutations and polymorphisms have been linked to several neurological disorders, including Alzheimer s disease (AD), frontotemporal dementia (FTD), Parkinson s disease (PD), and Nasu-Hakola disease. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhao, Y., et al. (2018). Neuron. 97(5); 1023-1031; Ulland, TK., et al. (2017). Cell. 170(4); 649-663; Yeh, FL., et al. (2016). Neuron. 91(2); 328-340).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service