Skip to Content
Merck
All Photos(12)

Documents

ZRB1246

Sigma-Aldrich

Anti-Mn-SOD Antibody, clone 1C14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C14, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~24.75 kDa
observed mol wt ~25 kDa

species reactivity

rat, human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C14 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects human Mn-SOD.

Immunogen

His-tagged full-length mature (without the transit peptide) recombinant human Mn-SOD.

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Mn-SOD in HepG2 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Mn-SOD in HeLa cell lysate.

Affinity Binding Assay:: A representative lot of this antibody bound Mn-SOD proteins with a KD of 4.5 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Mn-SOD in human colon tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Mn-SOD in PC12 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Mn-SOD, clone 1C14 ZooMAb®, Cat. No. ZRB1246, is a recombinant Rabbit monoclonal antibody that specifically targets Mn-SOD and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Superoxide dismutase [Mn], mitochondrial (UniProt: P04179; also known as EC:1.15.1.1, Mn-SOD) is encoded by the SOD2 gene (Gene ID: 6648) in human. Superoxide dismutases (SODs) provide defense against reactive oxygen species. Three types of SODs are found in organisms and are named based on the nature of the metal ion(s) found at their active sites. The Cu/Zn superoxide dismutase (Cu/ZnSOD), is primarily localized in the cytosol, nucleus, peroxisome, and intermembrane of mitochondria in eukaryotes. The Mn-SOD is found in the mitochondria, and the Fe-SOD is found primarily in bacteria. Mn-SOD is a homotetrameric enzyme, localized in the mitochondrial matrix, catalyzes the dismutation of superoxide radicals in mitochondria by converting anion superoxide into hydrogen peroxide and oxygen. Each monomer of Mn-SOD contains an active site bound to manganese (Mn) ion. Mn-SOD constitutes about 10 15% of the total SODs. The Mn ions can change between its 3+ and 2+ oxidation states within the dismutase mechanism that consists of two half-reactions. Electron exchange in metal complexes is reported to occur by either an inner-sphere or outer-sphere mechanism. In inner-sphere electron transfer, the metal cofactor is oxidized or reduced by a ligand bound directly to the metal, whereas in the outer-sphere mechanism, the metal is oxidized or reduced by a group not directly coordinated to the metal cofactor. The catalysis of superoxide anions by Mn-SOD occurs by an inner-sphere mechanism. Peroxynitrite radical can react with Mn-SOD that can cause a site-specific nitration of the critical tyrosine leading to its inactivation. Four isoforms of Mn-SOD have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Candas, D., and Li, JJ. (2014). Antioxid. Redox Signal. 20(10); 1599-1617).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service