Skip to Content
Merck
All Photos(20)

Key Documents

ZRB1169

Sigma-Aldrich

Anti-alpha Actinin 2 Antibody, clone 1E11, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-CMD1AA, Anti-CMH23, Anti-MPD6, Anti-MYOCOZ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E11, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 103.85 kDa
observed mol wt ~103 kDa

species reactivity

human, mouse

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ACTN2(88)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1E11 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Alpha-Actinin-2. It targets an epitope within 17 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to the first 17 amino acids from the N-terminal region of Alpha-actinin-2 from human skeletal muscle.

Application

Anti-alpha Actinin 2, clone 1E11 ZooMAb , Cat. No. ZRB1169, is a highly specific rabbit recombinant monoclonal antibody that targets alpha-Actinin-2 and has been tested for use in Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected alpha-Actinin-2 in mouse skeletal muscle tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected alpha- Actinin-2 in human skeletal muscle and mouse skeletal muscle tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected alpha-Actinin-2 in human skeletal muscle cells (SkMC).

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected alpha-Actinin-2 in human skeletal muscle frozen tissue sections.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Alpha-actinin-2 (UniProt: P35609; also known as Alpha-actinin skeletal muscle isoform 2, F-actin cross-linking protein) is encoded by the ACTN2 gene (Gene ID: 88) in human. Alpha-actinin-2 is expressed in both skeletal and cardiac muscle. It is considered as a bundling protein that acts to crosslink filamentous actin molecules and titin molecules from adjoining sarcomeres at Z-discs, a function that is modulated by phospholipids. It has a rod-shaped structure and it homodimerizes in an anti-parallel fashion. Each monomer contains an N-terminal actin-binding region composed of two calponin homology domains (aa 38-142 and 151-257), two C-terminal EF hand domains (aa 753-788 and 789-824), and four tandem spectrin-like repeats (aa 281-391, 401-506, 516-627, and 637-740). It has also been shown to localize to dendritic spines, enriched within the post-synaptic density (PSD). In rat hippocampal neurons loss of alpha-actinin-2 is shown to create an increased density of immature, filopodia-like protrusions that fail to mature into a mushroom-shaped spine during development and its knockdown prevents the recruitment and stabilization of PSD in the spine, resulting in failure of synapse formation. Mutations in ACTN2 gene is known to cause cardiomyopathy with ventricular hypertrophy leading to dyspnea, syncope, collapse, palpitations, and chest pain. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hodges, JL, et al. (2014). PLoS ONE 9(7): e101770).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingxuan Guo et al.
iScience, 27(6), 109954-109954 (2024-06-03)
Hypertension is a major cause of morbidity and mortality in patients with hypertrophic cardiomyopathy (HCM), suggesting a potential role for mechanics in HCM pathogenesis. Here, we developed an in vitro physiological model to investigate how mechanics acts together with HCM-linked myosin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service