Key Documents
Z768863
Tube clip for Eppendorf Tube®
secures lid for boiling
About This Item
UNSPSC Code:
41121702
NACRES:
NB.11
feature
fits 5 mL tubes
packaging
pkg of 10 ea
manufacturer/tradename
Eppendorf® 0030 119.509 (European)
Eppendorf® 0030119509 (US)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)