Key Documents
Z408514
Sigma-Aldrich
Scienceware® wash bottle carrier
Holds 6 x 8/16 oz bottles
Synonym(s):
bottle carriers, bottle holder
Select a Size
1 EA
$73.90
$73.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$73.90
About This Item
UNSPSC Code:
41122800
NACRES:
NB.17
$73.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
feature
Holds 6 x 8/16 oz bottles
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F11640-0001
L × W × H
286 mm × 194 mm × 225 mm
General description
Lightweight, PP carrier transports up to six 8 or 16 oz wash bottles. Autoclavable at 121°C. Bottles are not included.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)