Z768472
The Belly Button® mini-orbital shaker
with silicone mat, AC/DC input 90 - 240 V AC (universal plug)
Synonym(s):
amplification, cell culture growth, gel staining, immunoprecipitation, membrane rinsing, northern hybridization, phage elution, southern hybridization, western blotting
About This Item
Recommended Products
feature
0 ‑ 8 ° platform pitch control
manufacturer/tradename
IBI BBUAAUV1S
parameter
0-100 rpm speed
W × D × H
8 in. × 8 in. × 5 in.
platform W × L
7 in. × 7 in.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The Belly Button is an exceptionally simple liquid agitation system, consisting of orbiting platform flexibly anchored at four corners so that the platform motion, when adjusted by the platform pitch control collar, is an extremely effective combination of rocking platform and orbital shaker. One-dimensional shakers do not have the versatility and range of application that result from the variable speed and adjustable platform pitch of the Belly Button.
Ideal to use in a crowded incubator for call culture and tissue culture applications.
To use this product in the UK please order Z768588 separately
Legal Information
required but not provided
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service