Key Documents
Z765597
Tube rack for Benchmixer XL multi-tube vortexer
Holds 15 x 50 mL tubes
Select a Size
1 EA
$99.40
$99.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$99.40
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
$99.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
Holds 15 x 50 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific BV1010-500
General description
Benchmixer XL vortexer head adapters to hold multiple tubes.
This rack enables the use of 15 x 50 mL tubes in the BenchMixer XL heavy-duty hands-free mixing vortex mixer. It is ideal for the QuEChERS pesticide analysis method.
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)