Skip to Content
Merck

Skip To

C8358

Cube Rack

assorted colors

Synonym(s):

tube rack

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 ea
Check Cart for Availability
$157.00

About This Item

NACRES:
NB.35
UNSPSC Code:
41122804
eCl@ss:
32032201
Manufacturer/tradename:
Heathrow Scientific HS29050A
Feature:
Holds 16 x 11.5 mm tubes , Holds 10 x 15 mL tubes, Holds 10 x 17 mm tubes, Holds 12 x 12.5 mm tubes, Holds 12 x 3-7 mL tubes, Holds 16 x 1.5-2 mL tubes, Holds 4 x 30 mm tubes, Holds 4 x 50 mL tubes
Material:
polypropylene
Packaging:
pack of 5 ea

$157.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

description

Autoclavable

feature

Holds 16 x 11.5 mm tubes , Holds 10 x 15 mL tubes, Holds 10 x 17 mm tubes, Holds 12 x 12.5 mm tubes, Holds 12 x 3-7 mL tubes, Holds 16 x 1.5-2 mL tubes, Holds 4 x 30 mm tubes, Holds 4 x 50 mL tubes

packaging

pack of 5 ea

manufacturer/tradename

Heathrow Scientific HS29050A

size

108x113x113 mm

color

assorted colors

General description

Interlocking on all sides! Each one accommodates (4) 50 mL tubes, (10) 15 mL conical tubes, (12) 12 × 75 or 12 × 100 tubes, or (16) 1.5-2.0 mL microcentrifuge tubes. They slide together on any side allowing the user to work with different tubes on as many connected racks as desired.

Application

General purpose racks to hold a wide variety of tubes

Features and Benefits

  • Each side of the rack holds a differnet size tube maximizing versatility
  • Each rack is interlocking so that you can form a customised, sturdy tube holder to suit your needs

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HS120686R5651Z675970
description

assorted colors

description

green

description

assorted colors

description

Large tubes

feature

Holds 10 x 15 mL tubes, Holds 4 x 30 mm tubes, Holds 16 x 1.5-2 mL tubes, Holds 16 x 11.5 mm tubes , Holds 12 x 12.5 mm tubes, Holds 4 x 50 mL tubes, Holds 10 x 17 mm tubes, Holds 12 x 3-7 mL tubes

feature

to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes

feature

Holds 32 x 0.5 mL tubes, Holds 32 x 1.5-2 mL tubes, Holds 12 x 15 mL tubes, Holds 4 x 50 mL tubes

feature

Large tubes, Holds 2 x 29 mm tubes, Holds 8 x 20 mm tubes, Holds 8 x 10-15 mL tubes, Holds 10 x 10-13 mL tubes, Holds 2 x 50 mL tubes, Holds 10 x 13.5 mm tubes, Holds 6 x 15-20 mL tubes, Holds 6 x 16 mm tubes

manufacturer/tradename

Heathrow Scientific HS29050A

manufacturer/tradename

Heathrow Scientific 120686

manufacturer/tradename

Heathrow Scientific HS29022G

manufacturer/tradename

Heathrow Scientific HS29040A

material

polypropylene

material

green , polypropylene

material

polypropylene (heavy duty)

material

polypropylene

packaging

pack of 5 ea

packaging

pack of 5 ea

packaging

-

packaging

-

description

Autoclavable

description

-

description

Autoclavable

description

-

size

108x113x113 mm

size

-

size

174x95x52 mm

size

224x110x110 mm




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library