Key Documents
Z744064
FoilSeal RollMate Dispenser with 1 Roll of FoilSeal Film
case of 1 ea (500 Pre-Scored Sheets)
Select a Size
1 EACH
$509.00
$509.00
Estimated to ship onMay 10, 2025
Select a Size
Change View
1 EACH
$509.00
About This Item
UNSPSC Code:
41122410
NACRES:
NC.01
$509.00
Estimated to ship onMay 10, 2025
feature
RNase and DNase free
packaging
case of 1 ea (500 Pre-Scored Sheets)
manufacturer/tradename
Excel Scientific F2-RL-500
parameter
-20 °C temp. range (120 °C)
General description
  • An economical, efficient way to rapidly seal microplates; faster than applying single sheeted film
  • Self-standing, non-slip supports hold pre-scored film roll securely in place for quick and easy dispensing
  • Continuous backing liner can be torn off or fed directly into a waste basket for disposal
  • Roll tension can be easily adjusted to user preference (e.g. loose, tight)
  • SealPlate film minimizes evaporation, reduces cross-contamination and prevents spillage
  • 2.0 mil aluminum foil removes easily without tearing
  • Eliminates the “edge effect”, excess evaporation in the outside wells, which can occur with other films and plate lids
  • Useful as a simple cover to protect samples from external contamination, dust and debris
  • High integrity packaging prevents damage during transit; poly bag eliminates dust and particulate contamination
  • Pierceable with a pipette tip or robotic probe
  • Not recommended for long term cold storage
Legal Information
FoilSeal is a trademark of Excel Scientific, Inc.
RollMate is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)