Skip to Content
Merck
All Photos(1)

Key Documents

Z183423

Foil/poly bag

size 32 in. × 24 in.

Synonym(s):

Foil Packaging, Packaging Bag, Polyethylene Bag

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$84.30

$84.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 PKG
$84.30

About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

$84.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pkg of 5 ea

size

32 in. × 24 in.

thickness

5 mil

Looking for similar products? Visit Product Comparison Guide

General description

Heat sealable. Useful for storing air, light, or moisture-sensitive reagents, odoriferous products, and for temporarily containing leaking bottles.

Recommended heat seal conditions: 400°F (204°C), 40 psi, 1.5 sec.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can the bags be heat sealed? or what device do I need to seal them?

    1 answer
    1. Item Z183423 is heat sealable at 350°F (178°C). There are several sealers recommended for these bags. These sealers differ only by plug type. See the link below to review these options:
      https://www.sigmaaldrich.com/US/en/substance/bagheatsealerwithcutter1234598765

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service