Key Documents
Z743999
Worthington Accessories
necktube core for LS6000 liquid nitrogen refrigerator
Select a Size
1 EA
$934.00
$934.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$934.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$934.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Worthington RS60-9C16
technique(s)
cryopreservation: suitable
General description
All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.
Choose from one of the most recent versions:
Certificates of Analysis (COA)