Z743609
Tough-Spots®
diam. 3/8 in., pink, roll of 1000
Synonym(s):
solvent resistant labels, temperature resistant labels
Select a Size
$65.50
Select a Size
About This Item
$65.50
Recommended Products
packaging
roll of 1000
manufacturer/tradename
Diversified Biotech T-SPOTS-P
technique(s)
PCR: suitable
diam.
3/8 in.
color
pink
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Temperature-Resistant - From -196°C to 100°C, Tough-Spots® withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
Specially Designed - Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
Ideally Sized - Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Color Coding - 3/8 in. (9.5 mm) and 1/2 in. (13 mm) diameter Laser Tough-Spots® are available in White, Blue, Green, Orange, Red and Yellow
Features and Benefits
- Chemically inert, polyolefin labels resist most organic solvents and caustic agents.
- They adhere to most plastics†, glass, and metals without cracking, peeling, or degrading
- Temperature-Resistant from -196°C to 100°C
- Withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
- Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
- Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service