Z743599
Tough-Spots®
diam. 3/8 in., caramel, roll of 1000
Synonym(s):
solvent resistant labels, temperature resistant labels
Select a Size
$65.50
Estimated to ship onApril 25, 2025
Select a Size
About This Item
$65.50
Estimated to ship onApril 25, 2025
Recommended Products
packaging
roll of 1000
manufacturer/tradename
Diversified Biotech T-SPOTS-C
technique(s)
PCR: suitable
diam.
3/8 in.
color
caramel
Looking for similar products? Visit Product Comparison Guide
General description
Temperature-Resistant - From -196°C to 100°C, Tough-Spots® withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
Specially Designed - Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
Ideally Sized - Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Color Coding - 3/8 in. (9.5 mm) and 1/2 in. (13 mm) diameter Laser Tough-Spots® are available in White, Blue, Green, Orange, Red and Yellow
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service