Key Documents
Z741750
3-channel flow cell
Cell Only, γ-irradiated, Sealed
Select a Size
1 EA
$187.00
$187.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$187.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
$187.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
sterility
γ-irradiated
manufacturer/tradename
IBI ACCFL0001
General description
The IBI 3-channel Flow Cell (formerly Stovall 3-channel Flow Cell) is designed as a one-time use, affordable, sterile 3-channel flow cell apparatus for the creation of biofilms. This device allows for real-time, direct, non-destructive, on-line examination of biofilms with either a traditional transmission light microscope or a scanning confocal laser microscope which will produce a 3-D image of the biofilm.
The 3-channel flow cell is gamma irradiated and comes complete with bubble trap (ACCFL0002) and peristaltic pump tubing and connective tubing. The 3-channel flow cell contains a #1 Glass cover slip. The underside of this cover slip is where the biofilm will attach and grow. When the experiment is complete, the cover slip is cut using a hand-held glass cutter (ACCFL0014) and removed along with the biofilm.
Features and Benefits
  • The 3 separate channels allow for the simultaneous growth of 3 individual biofilms in the same cell.
  • 3-channel cell only – No tubing included.
  • Cell comes sterilized (gamma irradiated) and ready for use.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Bora Shin et al.
Applied microbiology and biotechnology, 104(3), 1259-1271 (2019-12-22)
The genomes of several Acinetobacter species possess three distinct polysaccharide-producing operons [two poly-N-acetyl glucosamine (PNAG) and one K-locus]. Using a microfluidic device, an increased amount of polysaccharides and enhanced biofilm formation were observed following continuous exposure to H2O2 and removal