Key Documents
Z741758
4-liter waste bottle
Select a Size
1 EA
$754.00
$754.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$754.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
$754.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
material
polycarbonate
feature
Comes with cap, tubing and fixtures.
manufacturer/tradename
IBI ACCFL0009
General description
The 4L waste media bottle is set up to capture waste effluent from the IBI Flow Cell system. The bottle is made of polycarbonate and can be autoclaved. It comes complete with cap, containing fixtures and tubing to connect to the 3-channel or Convertible flow cell systems. Autoclavable & re-usable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)