Key Documents
Z742311
Accuris E2 Grade Calibration Weight Set
Select a Size
1 EA
$1,410.00
$1,410.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$1,410.00
About This Item
UNSPSC Code:
41111500
NACRES:
NB.71
$1,410.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
manufacturer/tradename
Benchmark Scientific W1101-6-100
Related Categories
General description
Premium Class E2 Weight Set is ideal for calibration of analytical balances with a 0.0001g readability. Precision manufactured from high purity, 316 stainless steel, these weights are corrosion resistant with magnetic susceptibility <0.005. The set includes 6 weights, 100mg to 100g, for checking repeatability, cornerload, linearity and span. The weights are supplied in a protective case with tweezers, a cleaning cloth and a certificate of verification.
Packaging
6 pcs: 1x100 mg, 1 x 10 g, 2 x 20 g, 1 x 50 g, 1 x 100 g, includes protective case, tweezers, dust brush and cleaning cloth
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)