Skip to Content
Merck
All Photos(1)

Documents

Z724297

Hettich® fixed angle rotor, 12 place

for UNIVERSAL 320/320R Centrifuge, Fixed Angle Rotor, 12 place

Synonym(s):

Hettich® centrifuge rotors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.22

feature

to hold 12 x 15 ml tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1615

parameter

16,582 max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models Universal 320/320R

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service